Assay Details
Target Gene Details
Entrez Gene ID: | 10474 |
Gene Name: | transcriptional adaptor 3 |
Gene Aliases: |
ADA3, NGG1, STAF54, TADA3L, hADA3 |
Location: |
Chr.3:9779964-9793011 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TADA3 | NM_001278270.1 | NP_001265199.1 | ||
| NM_006354.3 | 1 | 644 | NP_006345.1 | |
| NM_133480.2 | 1 | 644 | NP_597814.1 | |
| NR_103488.1 | 1 | 644 | ||
| AK000228.1 | 1 | 48 | ||
| AK292264.1 | ||||
| AL117487.1 | 1 | 307 | CAB55957.1 | |
| BC009240.2 | 1 | 369 | AAH09240.1 | |
| BC013433.1 | 1 | 307 | AAH13433.1 | |
| BM560517.1 | 1 | 426 | ||
| DB032623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map