Assay Details
Target Gene Details
Entrez Gene ID: | 91419 |
Gene Name: | ATP23 metallopeptidase and ATP synthase assembly factor homolog |
Gene Aliases: |
KUB3, XRCC6BP1 |
Location: |
Chr.12:57941504-57957269 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP23 | NM_001320408.1 | NP_001307337.1 | ||
| NM_001320409.1 | NP_001307338.1 | |||
| NM_001320410.1 | NP_001307339.1 | |||
| NM_033276.3 | NP_150592.1 | |||
| XM_017020206.1 | XP_016875695.1 | |||
| AF078164.2 | AAD31085.1 | |||
| BC012776.2 | ||||
| BC115381.1 | AAI15382.1 | |||
| BC115382.1 | AAI15383.1 | |||
| BM544500.1 | ||||
| BM557436.1 | ||||
| BU601924.1 | ||||
| DA711361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559046 | Chr.12:57768302 - 58084815 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map