Assay Details
Target Gene Details
Entrez Gene ID: | 55055 |
Gene Name: | zwilch kinetochore protein |
Gene Aliases: |
KNTC1AP, hZwilch |
Location: |
Chr.15:66505083-66549485 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZWILCH | NM_001287821.1 | NP_001274750.1 | ||
| NM_001287822.1 | NP_001274751.1 | |||
| NM_001287823.1 | NP_001274752.1 | |||
| NM_017975.4 | NP_060445.3 | |||
| AK000898.1 | ||||
| AK023175.1 | BAB14446.1 | |||
| AK027468.1 | BAB55133.1 | |||
| AK122787.1 | ||||
| AK291048.1 | ||||
| BC036900.1 | AAH36900.1 | |||
| BC090041.1 | AAH90041.1 | |||
| BX640701.1 | CAE45821.1 |
More Information
Set Membership: |
|

Genomic Map