Assay Details
Target Gene Details
Entrez Gene ID: | 4084 |
Gene Name: | MAX dimerization protein 1 |
Gene Aliases: |
BHLHC58, MAD, MAD1 |
Location: |
Chr.2:69915041-69942945 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXD1 | NM_001202513.1 | 6 | 2021 | NP_001189442.1 |
| NM_001202514.1 | 5 | 1994 | NP_001189443.1 | |
| NM_002357.3 | 6 | 2024 | NP_002348.1 | |
| BC098396.1 | 6 | 1994 | AAH98396.1 |
More Information
Set Membership: |
|

Genomic Map