Assay Details
Target Gene Details
Entrez Gene ID: | 102723704 |
Gene Name: | uncharacterized LOC102723704 |
Gene Aliases: |
- |
Location: |
Chr.4:102777037-102799575 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723704 | NR_125932.1 | |||
| AK098632.1 |
Target Gene Details
Entrez Gene ID: | 7323 |
Gene Name: | ubiquitin conjugating enzyme E2 D3 |
Gene Aliases: |
E2(17)KB3, UBC4/5, UBCH5C |
Location: |
Chr.4:102794383-102868893 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2D3 | NM_001300795.1 | 8 | 1896 | NP_001287724.1 |
| NM_003340.6 | 8 | 2203 | NP_003331.1 | |
| NM_181886.3 | 8 | 2411 | NP_871615.1 | |
| NM_181887.2 | 8 | 2181 | NP_871616.1 | |
| NM_181888.3 | 8 | 2060 | NP_871617.1 | |
| NM_181889.2 | 8 | 2083 | NP_871618.1 | |
| NM_181890.2 | 8 | 2025 | NP_871619.1 | |
| NM_181891.2 | 8 | 2368 | NP_871620.1 | |
| NM_181892.3 | 8 | 2443 | NP_871621.1 | |
| NM_181893.2 | 7 | 1873 | NP_871622.1 | |
| XM_017008584.1 | 7 | 2235 | XP_016864073.1 | |
| AI651378.1 | ||||
| AK095822.1 | BAC04632.1 | |||
| AK127304.1 | ||||
| AL050186.1 | ||||
| BC037894.2 | 8 | 2180 | AAH37894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526256 | Chr.4:102791432 - 102806853 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map