Assay Details
Target Gene Details
Entrez Gene ID: | 6886 |
Gene Name: | TAL bHLH transcription factor 1, erythroid differentiation factor |
Gene Aliases: |
SCL, TCL5, bHLHa17, tal-1 |
Location: |
Chr.1:47216290-47232389 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAL1 | NM_001287347.2 | 5 | 2358 | NP_001274276.1 |
| NM_001290403.1 | 5 | 2468 | NP_001277332.1 | |
| NM_001290404.1 | 6 | 2473 | NP_001277333.1 | |
| NM_001290405.1 | 5 | 2341 | NP_001277334.1 | |
| NM_001290406.1 | 3 | 1865 | NP_001277335.1 | |
| NM_003189.5 | 6 | 2586 | NP_003180.1 | |
| XM_005271160.4 | 5 | 3968 | XP_005271217.1 | |
| XM_017002187.1 | 5 | 3383 | XP_016857676.1 | |
| XM_017002188.1 | 5 | 2575 | XP_016857677.1 | |
| XM_017002189.1 | 5 | 2378 | XP_016857678.1 | |
| XM_017002190.1 | 5 | 3074 | XP_016857679.1 | |
| XM_017002191.1 | 4 | 2199 | XP_016857680.1 | |
| XM_017002193.1 | 4 | 1935 | XP_016857682.1 | |
| M61108.1 | 4 | 2719 | AAA36600.1 | |
| S53245.1 | 5 | 2358 | AAB19683.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116299558] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map