Assay Details
Target Gene Details
Entrez Gene ID: | 10395 |
Gene Name: | DLC1 Rho GTPase activating protein |
Gene Aliases: |
ARHGAP7, HP, STARD12, p122-RhoGAP |
Location: |
Chr.8:13083361-13604606 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLC1 | NM_001164271.1 | 14 | 4628 | NP_001157743.1 |
| NM_001316668.1 | 14 | 4816 | NP_001303597.1 | |
| NM_006094.4 | 14 | 4886 | NP_006085.2 | |
| NM_182643.2 | 18 | 6321 | NP_872584.2 | |
| XM_005273374.1 | 18 | 6202 | XP_005273431.1 | |
| XM_017012951.1 | 18 | 6072 | XP_016868440.1 | |
| XM_017012952.1 | 14 | 4619 | XP_016868441.1 | |
| AB051510.1 | 18 | 6207 | ||
| AK023128.1 | 1 | 831 | ||
| AL117604.1 | 1 | 423 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022834 | Chr.8:13067043 - 14121780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map