Assay Details
Target Gene Details
Entrez Gene ID: | 7203 |
Gene Name: | chaperonin containing TCP1 subunit 3 |
Gene Aliases: |
CCT-gamma, CCTG, PIG48, TCP-1-gamma, TRIC5 |
Location: |
Chr.1:156308961-156338415 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCT3 | NM_001008800.2 | 10 | 1471 | NP_001008800.1 |
| NM_005998.4 | 12 | 1585 | NP_005989.3 | |
| NR_036564.1 | 13 | 1747 | ||
| NR_036565.1 | 13 | 1698 | ||
| AB208882.1 | 12 | 1451 | BAD92119.1 | |
| AK126400.1 | 12 | 1372 | ||
| AK293477.1 | 10 | 1301 | ||
| AK300765.1 | 13 | 1579 | ||
| AK315923.1 | 11 | 1371 | ||
| AK316387.1 | 13 | 1490 | ||
| AL833197.1 | 12 | 1445 | CAI46192.1 | |
| AY524046.1 | 12 | 1410 | AAS94255.1 | |
| BC006501.2 | 12 | 1417 | ||
| BC008019.1 | 12 | 1427 | AAH08019.1 | |
| U17104.1 | 2 | 324 | AAC50068.1 | |
| X74801.1 | 12 | 1351 | CAA52808.1 |
More Information
Set Membership: |
|

Genomic Map