Assay Details
Target Gene Details
Entrez Gene ID: | 2104 |
Gene Name: | estrogen related receptor gamma |
Gene Aliases: |
ERR3, ERRgamma, NR3B3 |
Location: |
Chr.1:216503246-217137757 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549199 | Chr.1:216211566 - 216641360 on Build GRCh38 | Gain |
|
| nsv1011849 | Chr.1:216495039 - 216527821 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map