Assay Details
Target Gene Details
Entrez Gene ID: | 56267 |
Gene Name: | kynurenine aminotransferase 3 |
Gene Aliases: |
CCBL2, KAT3, KATIII |
Location: |
Chr.1:88935773-88992960 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KYAT3 | NM_001008661.2 | NP_001008661.1 | ||
| NM_001008662.2 | NP_001008662.1 | |||
| AF091090.1 | AAC72959.1 | |||
| AK057176.1 | ||||
| AK301326.1 | ||||
| AK315917.1 | ||||
| AL833593.1 | ||||
| AY028624.1 | AAK26163.1 | |||
| BC000819.1 | AAH00819.1 | |||
| CR450282.1 | CAG29278.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462472 | Chr.1:88885058 - 88971430 on Build GRCh38 | Loss |
|
| dgv207n100 | Chr.1:88932165 - 88995971 on Build GRCh38 | Loss |
|
| esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain |
|
| nsv1002798 | Chr.1:88954378 - 88988906 on Build GRCh38 | Gain |
|
| nsv830525 | Chr.1:88776863 - 88975647 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map