Assay Details
Target Gene Details
Entrez Gene ID: | 64801 |
Gene Name: | ARV1 homolog, fatty acid homeostasis modulator |
Gene Aliases: |
- |
Location: |
Chr.1:230979075-231000733 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARV1 | NM_022786.1 | 4 | 571 | NP_073623.1 |
| XM_017002106.1 | 5 | 665 | XP_016857595.1 | |
| AF271780.1 | 4 | 571 | AAG44791.1 | |
| AF290878.1 | 4 | 571 | AAG47671.1 | |
| AF321442.1 | 4 | 605 | AAK11180.1 | |
| AK293049.1 | 4 | 577 | ||
| AL122047.1 | CAB59183.3 | |||
| BC016309.1 | 4 | 566 | AAH16309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv1011066 | Chr.1:230940432 - 231242850 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map