Assay Details
Target Gene Details
Entrez Gene ID: | 137682 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 6 |
Gene Aliases: |
C8orf38 |
Location: |
Chr.8:94895271-95118496 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF6 | XM_011516835.2 | XP_011515137.1 | ||
| XM_017013028.1 | XP_016868517.1 | |||
| XM_017013030.1 | XP_016868519.1 | |||
| XM_017013031.1 | XP_016868520.1 | |||
| XM_017013032.1 | XP_016868521.1 | |||
| XM_017013035.1 | XP_016868524.1 | |||
| XM_017013036.1 | XP_016868525.1 | |||
| XM_017013039.1 | XP_016868528.1 |
Target Gene Details
Entrez Gene ID: | 94241 |
Gene Name: | tumor protein p53 inducible nuclear protein 1 |
Gene Aliases: |
SIP, TP53DINP1, TP53INP1A, TP53INP1B, Teap, p53DINP1 |
Location: |
Chr.8:94925972-94949387 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53INP1 | NM_001135733.1 | 5 | 5363 | NP_001129205.1 |
| NM_033285.3 | 4 | 5325 | NP_150601.1 | |
| XM_011517386.2 | 4 | 5177 | XP_011515688.1 | |
| AB017926.1 | 3 | 4980 | BAB62149.1 | |
| AB017927.1 | 4 | 5018 | BAB62150.1 | |
| AF409114.1 | 5 | 5335 | AAL04513.1 | |
| AF409115.1 | 4 | 5297 | AAL04514.1 | |
| AK025792.1 | 1 | 1749 | ||
| AK123608.1 | 1 | 3517 | ||
| AK125880.1 | 5 | 5342 | ||
| AK130074.1 | 1 | 1329 | ||
| AL133074.1 | 1 | 2562 | ||
| AL833652.1 | 4 | 4943 |
More Information
Set Membership: |
|

Genomic Map