Assay Details
Target Gene Details
Entrez Gene ID: | 84196 |
Gene Name: | ubiquitin specific peptidase 48 |
Gene Aliases: |
RAP1GA1, USP31 |
Location: |
Chr.1:21678298-21783620 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP48 | NM_032236.5 | 27 | 4260 | NP_115612.4 |
| AI248244.1 | 1 | 206 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34155 | Chr.1:21367084 - 21840123 on Build GRCh38 | Loss |
|
| nsv834391 | Chr.1:21523440 - 21723989 on Build GRCh38 | Loss |
|
| nsv5854 | Chr.1:21676656 - 21711616 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map