Assay Details
Target Gene Details
Entrez Gene ID: | 883 |
Gene Name: | kynurenine aminotransferase 1 |
Gene Aliases: |
CCBL1, GTK, KAT1, KATI |
Location: |
Chr.9:128832942-128882551 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KYAT1 | NM_001122671.1 | 7 | 827 | NP_001116143.1 |
| NM_001122672.1 | 6 | 683 | NP_001116144.1 | |
| NM_001287390.1 | 9 | 1224 | NP_001274319.1 | |
| NM_004059.4 | 7 | 833 | NP_004050.3 | |
| NR_109829.1 | 7 | 833 | ||
| XM_006717311.3 | 8 | 1019 | XP_006717374.1 | |
| XM_011519165.2 | 9 | 1195 | XP_011517467.1 | |
| XM_011519167.2 | 7 | 799 | XP_011517469.2 | |
| XM_011519168.1 | 6 | 658 | XP_011517470.1 | |
| XM_011519169.1 | 7 | 927 | XP_011517471.1 | |
| XM_011519170.2 | 7 | 1295 | XP_011517472.1 | |
| XM_017015260.1 | 8 | 979 | XP_016870749.1 | |
| XM_017015261.1 | 9 | 1201 | XP_016870750.1 | |
| XM_017015262.1 | 9 | 1690 | XP_016870751.1 | |
| XM_017015263.1 | 9 | 1105 | XP_016870752.1 | |
| XM_017015264.1 | 8 | 1012 | XP_016870753.1 | |
| XM_017015265.1 | 7 | 1389 | XP_016870754.1 | |
| XM_017015266.1 | 8 | 1020 | XP_016870755.1 | |
| XM_017015267.1 | 7 | 841 | XP_016870756.1 | |
| XM_017015268.1 | 8 | 1003 | XP_016870757.1 | |
| AK094505.1 | 7 | 1376 | ||
| AK291178.1 | 7 | 821 | ||
| AK297995.1 | 9 | 1080 | ||
| AK314427.1 | 7 | 703 | ||
| BC033685.1 | 6 | 537 | AAH33685.1 | |
| BI755163.1 | 6 | 537 | ||
| EL733697.1 | 5 | 498 | ||
| X82224.1 | 6 | 648 | CAA57702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6724 | Chr.9:128823206 - 128858415 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map