Assay Details
Target Gene Details
Entrez Gene ID: | 11128 |
Gene Name: | RNA polymerase III subunit A |
Gene Aliases: |
ADDH, HLD7, RPC1, RPC155, hRPC155 |
Location: |
Chr.10:77975149-78029540 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3A | NM_007055.3 | 27 | 3633 | NP_008986.2 |
| AF021351.1 | 28 | 3500 | AAB86536.1 | |
| AK026023.1 | 18 | 2207 | ||
| AK300329.1 | 28 | 3550 | ||
| AY091461.1 | AAM12031.1 | |||
| AY091462.1 | 3 | 227 | AAM12032.1 | |
| BC014399.1 | 4 | 339 | AAH14399.1 | |
| BC041089.1 | 27 | 3594 | AAH41089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7310 | Chr.10:77962983 - 78007707 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map