Assay Details
Target Gene Details
Entrez Gene ID: | 84441 |
Gene Name: | mastermind like transcriptional coactivator 2 |
Gene Aliases: |
MAM-3, MAM2, MAM3, MLL-MAML2 |
Location: |
Chr.11:95976593-96343245 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAML2 | NM_032427.3 | 5 | 4172 | NP_115803.1 |
| XM_011543023.2 | 5 | 6007 | XP_011541325.1 | |
| XM_011543024.2 | 5 | 2712 | XP_011541326.1 | |
| AB058722.1 | 5 | 4163 | ||
| AK308157.1 | 6 | 3939 | ||
| AY040322.1 | 5 | 4163 | AAK93831.1 | |
| BC143529.1 | 5 | 2913 | ||
| BC152449.1 | 5 | 4163 | ||
| CR627398.1 | 1 | 3168 | CAH10491.1 |
More Information
Set Membership: |
|

Genomic Map