Assay Details
Target Gene Details
Entrez Gene ID: | 338599 |
Gene Name: | dual specificity phosphatase and pro isomerase domain containing 1 |
Gene Aliases: |
DUSP27, FMDSP |
Location: |
Chr.10:75026427-75073827 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUPD1 | NM_001003892.1 | NP_001003892.1 | ||
| XM_011539747.2 | 5 | 1117 | XP_011538049.1 | |
| AY686755.1 | AAT94288.1 | |||
| BC137321.1 | ||||
| BC137322.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv926n100 | Chr.10:74994872 - 75081593 on Build GRCh38 | Gain |
|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map