Assay Details
Target Gene Details
Entrez Gene ID: | 80021 |
Gene Name: | transmembrane protein 62 |
Gene Aliases: |
- |
Location: |
Chr.15:43133115-43185147 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM62 | NM_024956.3 | 5 | 813 | NP_079232.3 |
| XM_005254682.4 | 4 | 780 | XP_005254739.1 | |
| XM_005254683.4 | 3 | 669 | XP_005254740.1 | |
| XM_005254684.3 | XP_005254741.1 | |||
| XM_011522071.1 | 4 | 297 | XP_011520373.1 | |
| XM_011522072.2 | 3 | 464 | XP_011520374.1 | |
| XM_011522073.1 | XP_011520375.1 | |||
| XM_011522074.2 | XP_011520376.1 | |||
| XM_017022600.1 | 4 | 812 | XP_016878089.1 | |
| XM_017022601.1 | 4 | 565 | XP_016878090.1 | |
| XM_017022602.1 | 3 | 701 | XP_016878091.1 | |
| XM_017022603.1 | 4 | 297 | XP_016878092.1 | |
| XM_017022604.1 | 4 | 781 | XP_016878093.1 | |
| XM_017022605.1 | 3 | 670 | XP_016878094.1 | |
| XM_017022606.1 | XP_016878095.1 | |||
| XM_017022607.1 | XP_016878096.1 | |||
| XM_017022608.1 | XP_016878097.1 | |||
| XM_017022609.1 | XP_016878098.1 | |||
| XM_017022610.1 | XP_016878099.1 | |||
| XM_017022611.1 | XP_016878100.1 | |||
| XM_017022612.1 | XP_016878101.1 | |||
| XM_017022613.1 | XP_016878102.1 | |||
| XM_017022614.1 | XP_016878103.1 | |||
| XM_017022615.1 | XP_016878104.1 | |||
| XM_017022616.1 | XP_016878105.1 | |||
| AK027028.1 | 2 | 118 | BAB15630.1 | |
| AK225353.1 | 2 | 118 | ||
| AK309679.1 | 4 | 329 | ||
| BC009981.2 | 5 | 813 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525461 | Chr.15:42770350 - 43189929 on Build GRCh38 | Loss |
|
| nsv569241 | Chr.15:42908911 - 43230724 on Build GRCh38 | Loss |
|
| nsv515876 | Chr.15:42824708 - 43153107 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map