Assay Details
Target Gene Details
Entrez Gene ID: | 10059 |
Gene Name: | dynamin 1 like |
Gene Aliases: |
DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV |
Location: |
Chr.12:32679200-32745650 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNM1L | NM_001278463.1 | 13 | 1655 | NP_001265392.1 |
| NM_001278464.1 | 14 | 1694 | NP_001265393.1 | |
| NM_001278465.1 | 14 | 1694 | NP_001265394.1 | |
| NM_001278466.1 | 10 | 1251 | NP_001265395.1 | |
| NM_005690.4 | 13 | 1655 | NP_005681.2 | |
| NM_012062.4 | 13 | 1655 | NP_036192.2 | |
| NM_012063.3 | 13 | 1655 | NP_036193.2 | |
| XM_005253282.4 | 14 | 1666 | XP_005253339.1 | |
| XM_011520543.2 | 14 | 1666 | XP_011518845.1 | |
| XM_011520544.2 | 12 | 1372 | XP_011518846.1 | |
| XM_017018663.1 | 12 | 1372 | XP_016874152.1 | |
| XM_017018664.1 | 12 | 1372 | XP_016874153.1 | |
| XM_017018665.1 | 12 | 1372 | XP_016874154.1 | |
| AB006965.1 | 13 | 1526 | BAA22193.1 | |
| AB209070.1 | 14 | 1571 | BAD92307.1 | |
| AF000430.1 | 13 | 1563 | AAC23724.1 | |
| AF061795.1 | 13 | 1512 | AAC35283.1 | |
| AF151685.1 | 13 | 1512 | AAD39541.1 | |
| AK291094.1 | 13 | 1535 | ||
| AK294533.1 | 10 | 1131 | ||
| AK299926.1 | 14 | 1607 | ||
| AK302565.1 | 13 | 1655 | ||
| BC024590.1 | 13 | 1525 | AAH24590.1 | |
| BC107763.1 | 13 | 1539 | AAI07764.1 |
Target Gene Details
Entrez Gene ID: | 51067 |
Gene Name: | tyrosyl-tRNA synthetase 2 |
Gene Aliases: |
CGI-04, MLASA2, MT-TYRRS, TYRRS |
Location: |
Chr.12:32725247-32755961 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YARS2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1429n100 | Chr.12:32718926 - 32775899 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map