Assay Details
Target Gene Details
Entrez Gene ID: | 92922 |
Gene Name: | coiled-coil domain containing 102A |
Gene Aliases: |
- |
Location: |
Chr.16:57512178-57538305 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC102A | NM_033212.3 | 9 | 1972 | NP_149989.2 |
| XM_011523469.2 | 9 | 3252 | XP_011521771.1 | |
| BC004307.1 | 6 | 929 | AAH04307.1 | |
| BC008285.1 | 9 | 1951 | AAH08285.1 | |
| BC009941.2 | 9 | 1972 | AAH09941.1 | |
| CA449463.1 | 1 | 352 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
| nsv952683 | Chr.16:57497089 - 57536488 on Build GRCh38 | Deletion |
|
| nsv572723 | Chr.16:57497721 - 57535535 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map