Assay Details
Target Gene Details
Entrez Gene ID: | 8140 |
Gene Name: | solute carrier family 7 member 5 |
Gene Aliases: |
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1 |
Location: |
Chr.16:87830022-87869499 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A5 | NM_003486.5 | 10 | 4478 | NP_003477.4 |
| XM_006721286.3 | 10 | 4121 | XP_006721349.2 | |
| AB018009.1 | 10 | 4471 | BAA84648.1 | |
| AF104032.1 | 10 | 4472 | AAD20464.1 | |
| BC039692.2 | 10 | 4503 | AAH39692.1 | |
| BC042600.1 | 10 | 4472 | AAH42600.1 | |
| M80244.1 | 10 | 3918 | AAA35780.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059786 | Chr.16:87811419 - 88019505 on Build GRCh38 | Gain |
|
| nsv833322 | Chr.16:87715029 - 87884870 on Build GRCh38 | Loss |
|
| nsv1160441 | Chr.16:87761248 - 87867260 on Build GRCh38 | Deletion |
|
| esv3892935 | Chr.16:87808912 - 88010269 on Build GRCh38 | Loss |
|
| dgv3062n100 | Chr.16:87725235 - 87842257 on Build GRCh38 | Loss |
|
| nsv573557 | Chr.16:87808535 - 87854711 on Build GRCh38 | Loss |
|
| nsv1062946 | Chr.16:87635801 - 88019505 on Build GRCh38 | Gain |
|
| dgv36n68 | Chr.16:87801736 - 87983550 on Build GRCh38 | Loss |
|
| nsv573536 | Chr.16:87705252 - 87914409 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73251309] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map