Assay Details
Target Gene Details
Entrez Gene ID: | 50484 |
Gene Name: | ribonucleotide reductase regulatory TP53 inducible subunit M2B |
Gene Aliases: |
MTDPS8A, MTDPS8B, P53R2 |
Location: |
Chr.8:102204501-102239118 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRM2B | NM_001172477.1 | 8 | 1070 | NP_001165948.1 |
| NM_001172478.1 | 7 | 933 | NP_001165949.1 | |
| NM_015713.4 | 8 | 1089 | NP_056528.2 | |
| AB036063.1 | 8 | 1089 | BAA92434.1 | |
| AB163437.1 | 7 | 689 | BAD11774.1 | |
| AB163438.1 | 3 | 178 | BAD11775.1 | |
| AB166669.1 | BAD12265.1 | |||
| AB166670.1 | 3 | 209 | BAD12266.1 | |
| AB166671.1 | 8 | 845 | BAD12267.1 | |
| AK001965.1 | 8 | 884 | BAA92005.1 | |
| AK304354.1 | 8 | 1014 | ||
| AL137348.1 | 8 | 891 | CAB70703.2 | |
| BC042468.1 | 6 | 647 | AAH42468.1 | |
| BC108261.1 | 8 | 901 | AAI08262.1 | |
| BC117496.1 | 8 | 926 | ||
| BC130628.1 | 8 | 926 | ||
| BX509409.1 | 2 | 93 | ||
| CR627376.1 | 8 | 1014 | CAH10473.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572925 | Chr.8:102200769 - 102219567 on Build GRCh38 | Loss |
|
| esv3576598 | Chr.8:102200959 - 102574604 on Build GRCh38 | Gain |
|
| nsv1025935 | Chr.8:102186979 - 102456094 on Build GRCh38 | Gain |
|
| esv3618218 | Chr.8:102200369 - 102319026 on Build GRCh38 | Gain |
|
| nsv1020359 | Chr.8:102097693 - 102306187 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map