Assay Details
Target Gene Details
Entrez Gene ID: | 268 |
Gene Name: | anti-Mullerian hormone |
Gene Aliases: |
MIF, MIS |
Location: |
Chr.19:2249114-2252073 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMH | NM_000479.3 | NP_000470.2 | ||
| BC049194.1 | AAH49194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578382 | Chr.19:2194897 - 2260362 on Build GRCh38 | Loss |
|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv578383 | Chr.19:2214058 - 2285197 on Build GRCh38 | Loss |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map