Assay Details
Target Gene Details
Entrez Gene ID: | 10974 |
Gene Name: | adipogenesis regulatory factor |
Gene Aliases: |
AFRO, APM2, C10orf116, apM-2 |
Location: |
Chr.10:86968431-86970909 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADIRF | NM_006829.2 | 3 | 585 | NP_006820.1 |
| BM850559.1 | ||||
| CD722742.1 | 1 | 195 |
Target Gene Details
Entrez Gene ID: | 119385 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 11 |
Gene Aliases: |
- |
Location: |
Chr.10:86970741-87010203 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP11 | NM_133447.1 | 1 | 82 | NP_597704.1 |
| AK097165.1 | 1 | 82 |
Target Gene Details
Entrez Gene ID: | 79812 |
Gene Name: | multimerin 2 |
Gene Aliases: |
EMILIN-3, EMILIN3, ENDOGLYX-1 |
Location: |
Chr.10:86935540-86971311 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMRN2 | XM_006717970.3 | 1 | 490 | XP_006718033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428236 | Chr.10:86866859 - 87550005 on Build GRCh38 | Loss |
|
| nsv517487 | Chr.10:86899290 - 86973435 on Build GRCh38 | Gain+Loss |
|
| esv2759774 | Chr.10:86755301 - 87550005 on Build GRCh38 | Gain+Loss |
|
| nsv470969 | Chr.10:86936624 - 86972907 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73334941] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map