Assay Details
Target Gene Details
Entrez Gene ID: | 1990 |
Gene Name: | chymotrypsin like elastase family member 1 |
Gene Aliases: |
ELA1 |
Location: |
Chr.12:51328443-51346679 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CELA1 | NM_001971.5 | 8 | 842 | NP_001962.3 |
| AF120493.1 | 8 | 842 | AAD28441.1 | |
| BC069454.1 | AAH69454.1 | |||
| BC075091.2 | AAH75091.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469377 | Chr.12:51296967 - 51340167 on Build GRCh38 | Loss |
|
| nsv832408 | Chr.12:51302821 - 51477555 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map