Assay Details
Target Gene Details
Entrez Gene ID: | 80854 |
Gene Name: | SET domain containing lysine methyltransferase 7 |
Gene Aliases: |
KMT7, SET7, SET7/9, SET9 |
Location: |
Chr.4:139495934-139556769 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD7 | NM_001306199.1 | NP_001293128.1 | ||
| NM_030648.3 | 8 | 7222 | NP_085151.1 | |
| XM_017008661.1 | 6 | 6315 | XP_016864150.1 | |
| AB051504.1 | 8 | 6857 | ||
| AK024846.1 | 1 | 1887 | ||
| AK310856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894040 | Chr.4:139499096 - 139522362 on Build GRCh38 | Loss |
|
| nsv830085 | Chr.4:139393507 - 139586168 on Build GRCh38 | Loss |
|
| nsv830086 | Chr.4:139494751 - 139671217 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map