Assay Details
Target Gene Details
Entrez Gene ID: | 10487 |
Gene Name: | adenylate cyclase associated protein 1 |
Gene Aliases: |
CAP, CAP1-PEN |
Location: |
Chr.1:40040070-40072649 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAP1 | NM_001105530.1 | NP_001099000.1 | ||
| NM_006367.3 | 1 | 160 | NP_006358.1 | |
| XM_005270368.1 | XP_005270425.1 | |||
| XM_011540509.1 | XP_011538811.1 | |||
| XM_011540510.1 | XP_011538812.1 | |||
| XM_011540511.1 | XP_011538813.1 | |||
| XM_011540512.1 | XP_011538814.1 | |||
| XM_011540513.2 | XP_011538815.1 | |||
| XM_011540514.1 | 1 | 48 | XP_011538816.1 | |
| XM_011540515.1 | XP_011538817.1 | |||
| XM_017000067.1 | 1 | 65 | XP_016855556.1 | |
| XM_017000068.1 | 1 | 44 | XP_016855557.1 | |
| XM_017000070.1 | XP_016855559.1 | |||
| XM_017000072.1 | XP_016855561.1 | |||
| XM_017000073.1 | 1 | 60 | XP_016855562.1 | |
| AK096193.1 | ||||
| AK222513.1 | BAD96233.1 | |||
| AK295396.1 | ||||
| AK297829.1 | ||||
| AK298094.1 | ||||
| AK298110.1 | ||||
| AK300861.1 | ||||
| AK315731.1 | ||||
| BC013963.2 | AAH13963.1 | |||
| BC095440.1 | AAH95440.1 | |||
| BI460362.1 | ||||
| M98474.1 | AAA35648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
| nsv950665 | Chr.1:40031829 - 40062428 on Build GRCh38 | Duplication |
|
| esv2750836 | Chr.1:40038107 - 40093576 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145869294] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map