Assay Details
Target Gene Details
Entrez Gene ID: | 23185 |
Gene Name: | La ribonucleoprotein domain family member 4B |
Gene Aliases: |
KIAA0217, LARP5 |
Location: |
Chr.10:806914-931900 on Build GRCh38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP4B | NM_015155.2 | NP_055970.1 | ||
| XM_005252432.3 | XP_005252489.1 | |||
| XM_005252435.3 | XP_005252492.1 | |||
| XM_011519435.2 | XP_011517737.1 | |||
| XM_017015987.1 | XP_016871476.1 | |||
| XM_017015988.1 | XP_016871477.1 | |||
| XM_017015989.1 | XP_016871478.1 | |||
| XM_017015990.1 | XP_016871479.1 | |||
| XM_017015992.1 | XP_016871481.1 | |||
| XM_017015993.1 | XP_016871482.1 | |||
| XM_017015994.1 | XP_016871483.1 | |||
| BC131630.1 | ||||
| BC152443.1 | ||||
| CR749653.1 | CAH18446.1 | |||
| D86971.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549753 | Chr.10:412537 - 848898 on Build GRCh38 | Gain |
|
| esv2660097 | Chr.10:526001 - 1246069 on Build GRCh38 | Deletion |
|
| nsv1048707 | Chr.10:788499 - 869594 on Build GRCh38 | Gain |
|
| nsv466698 | Chr.10:790175 - 842133 on Build GRCh38 | Loss |
|
| nsv549765 | Chr.10:688289 - 1083587 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map