Assay Details
Target Gene Details
Entrez Gene ID: | 10513 |
Gene Name: | amyloid beta precursor protein binding protein 2 |
Gene Aliases: |
APP-BP2, HS.84084, PAT1 |
Location: |
Chr.17:60443149-60526240 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APPBP2 | NM_001282476.1 | 12 | 5690 | NP_001269405.1 |
| NM_006380.3 | 13 | 5779 | NP_006371.2 | |
| XM_011524198.2 | 14 | 5506 | XP_011522500.1 | |
| XM_011524199.2 | 10 | 4991 | XP_011522501.1 | |
| D86981.1 | 13 | 5758 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3273n100 | Chr.17:60293516 - 60519251 on Build GRCh38 | Gain |
|
| esv3640926 | Chr.17:60440546 - 60447832 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map