Assay Details
Target Gene Details
Entrez Gene ID: | 51582 |
Gene Name: | antizyme inhibitor 1 |
Gene Aliases: |
AZI, AZIA1, OAZI, OAZIN, ODC1L |
Location: |
Chr.8:102826302-102864200 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AZIN1 | NM_001301668.1 | NP_001288597.1 | ||
| NM_015878.5 | NP_056962.2 | |||
| NM_148174.3 | NP_680479.1 | |||
| XM_005250969.1 | XP_005251026.1 | |||
| XM_011517126.1 | XP_011515428.1 | |||
| XM_011517127.1 | XP_011515429.1 | |||
| XM_011517128.2 | XP_011515430.1 | |||
| XM_017013563.1 | XP_016869052.1 | |||
| XM_017013564.1 | XP_016869053.1 | |||
| XM_017013565.1 | XP_016869054.1 | |||
| XM_017013566.1 | XP_016869055.1 | |||
| AK095493.1 | ||||
| AK290516.1 | ||||
| BC013420.2 | AAH13420.1 | |||
| BC019279.1 | AAH19279.1 | |||
| BT006858.1 | AAP35504.1 | |||
| CR456745.1 | CAG33026.1 | |||
| D88674.1 | BAA23593.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611822 | Chr.8:102804878 - 102882098 on Build GRCh38 | Loss |
|
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map