Assay Details
Target Gene Details
Entrez Gene ID: | 6886 |
Gene Name: | TAL bHLH transcription factor 1, erythroid differentiation factor |
Gene Aliases: |
SCL, TCL5, bHLHa17, tal-1 |
Location: |
Chr.1:47216290-47232389 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAL1 | NM_001287347.2 | 5 | 4459 | NP_001274276.1 |
| NM_001290403.1 | 5 | 4569 | NP_001277332.1 | |
| NM_001290404.1 | 6 | 4574 | NP_001277333.1 | |
| NM_001290405.1 | 5 | 4442 | NP_001277334.1 | |
| NM_001290406.1 | 3 | 3966 | NP_001277335.1 | |
| NM_003189.5 | 6 | 4687 | NP_003180.1 | |
| XM_005271160.4 | 5 | 6069 | XP_005271217.1 | |
| XM_017002187.1 | 5 | 5484 | XP_016857676.1 | |
| XM_017002188.1 | 5 | 4676 | XP_016857677.1 | |
| XM_017002189.1 | 5 | 4479 | XP_016857678.1 | |
| XM_017002190.1 | 5 | 5175 | XP_016857679.1 | |
| XM_017002191.1 | 4 | 4300 | XP_016857680.1 | |
| XM_017002193.1 | 4 | 4036 | XP_016857682.1 | |
| BQ186967.1 | 1 | 338 | ||
| M61108.1 | 4 | 4820 | AAA36600.1 | |
| N71056.1 | 1 | 199 | ||
| S53245.1 | 5 | 4459 | AAB19683.1 |
More Information
Set Membership: |
|

Genomic Map