Assay Details
Target Gene Details
Entrez Gene ID: | 83857 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 1 |
Gene Aliases: |
ARG99, OLF, TMTC1A |
Location: |
Chr.12:29500813-29784759 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMTC1 | NM_001193451.1 | 18 | 4661 | NP_001180380.1 |
| NM_175861.3 | 18 | 4752 | NP_787057.2 | |
| XM_005253499.4 | 20 | 4913 | XP_005253556.1 | |
| XM_017020003.1 | 19 | 4746 | XP_016875492.1 | |
| XM_017020004.1 | 19 | 4706 | XP_016875493.1 | |
| XM_017020005.1 | 17 | 4519 | XP_016875494.1 | |
| XM_017020007.1 | 16 | 3939 | XP_016875496.1 | |
| AL832328.1 | 4 | 2121 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557917 | Chr.12:29459492 - 29658538 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74583253] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map