Assay Details
Target Gene Details
Entrez Gene ID: | 257397 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 3 |
Gene Aliases: |
MAP3K7IP3, NAP1 |
Location: |
Chr.X:30827442-30889394 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAB3 | NM_152787.4 | 11 | 5102 | NP_690000.3 |
| XM_005274483.3 | 10 | 5015 | XP_005274540.2 | |
| XM_005274484.2 | 10 | 4997 | XP_005274541.2 | |
| AK096786.1 | 1 | 1552 | ||
| AK297274.1 |
More Information
Set Membership: |
|

Genomic Map