Assay Details
Target Gene Details
Entrez Gene ID: | 151050 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 like |
Gene Aliases: |
C2orf67, MSL1v2 |
Location: |
Chr.2:210021421-210171370 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANSL1L | NM_001307976.1 | 14 | 4155 | NP_001294905.1 |
| NM_152519.3 | 15 | 4281 | NP_689732.2 | |
| XM_005246328.4 | 15 | 4563 | XP_005246385.1 | |
| XM_005246329.4 | 15 | 4269 | XP_005246386.1 | |
| XM_005246330.4 | 15 | 4150 | XP_005246387.1 | |
| XM_005246332.4 | 14 | 4436 | XP_005246389.1 | |
| XM_011510706.2 | 15 | 4188 | XP_011509008.1 | |
| XM_011510707.2 | 15 | 4371 | XP_011509009.1 | |
| XM_011510710.2 | 14 | 3020 | XP_011509012.1 | |
| XM_017003430.1 | 15 | 4307 | XP_016858919.1 | |
| XM_017003431.1 | 11 | 6389 | XP_016858920.1 | |
| AA252480.1 | 1 | 434 | ||
| AK074441.1 | 14 | 3050 | BAB85085.1 | |
| BQ948513.1 | 1 | 175 | ||
| BU568066.1 | 1 | 217 |
Target Gene Details
Entrez Gene ID: | 6120 |
Gene Name: | ribulose-5-phosphate-3-epimerase |
Gene Aliases: |
RPE2-1 |
Location: |
Chr.2:210002565-210029156 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPE | NM_001278282.1 | 7 | 2881 | NP_001265211.1 |
| NM_001278283.1 | 8 | 2961 | NP_001265212.1 | |
| NM_001278285.1 | 5 | 2789 | NP_001265214.1 | |
| NM_001278286.1 | 6 | 2805 | NP_001265215.1 | |
| NM_001278288.1 | 5 | 2796 | NP_001265217.1 | |
| NM_001278289.1 | 7 | 2945 | NP_001265218.1 | |
| NM_001318926.1 | 7 | 2930 | NP_001305855.1 | |
| NM_001318927.1 | 7 | 2974 | NP_001305856.1 | |
| NM_001318928.1 | 7 | 2825 | NP_001305857.1 | |
| NM_001318929.1 | 6 | 2872 | NP_001305858.1 | |
| NM_001318930.1 | 6 | 2865 | NP_001305859.1 | |
| NM_001318931.1 | 6 | 2827 | NP_001305860.1 | |
| NM_006916.2 | 6 | 2850 | NP_008847.1 | |
| NM_199229.2 | 6 | 2876 | NP_954699.1 | |
| XM_006712677.3 | 7 | 2823 | XP_006712740.1 | |
| AB075523.1 | 1 | 1501 | ||
| AI222711.1 |
More Information
Set Membership: |
|

Genomic Map