Assay Details
Target Gene Details
Entrez Gene ID: | 94274 |
Gene Name: | protein phosphatase 1 regulatory inhibitor subunit 14A |
Gene Aliases: |
CPI-17, CPI17, PPP1INL |
Location: |
Chr.19:38251237-38256591 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R14A | NM_001243947.1 | 1 | 342 | NP_001230876.1 |
| NM_033256.2 | 1 | 342 | NP_150281.1 | |
| AB056508.1 | 1 | 283 | BAB62502.1 | |
| AB056509.1 | 1 | 283 | BAB62503.1 | |
| AF123763.1 | 1 | 133 | AAP97242.1 | |
| AY050670.1 | 1 | 127 | AAL25828.1 | |
| BC021089.1 | 1 | 107 | AAH21089.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833821 | Chr.19:38139297 - 38266152 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map