Assay Details
Target Gene Details
Entrez Gene ID: | 252969 |
Gene Name: | nei like DNA glycosylase 2 |
Gene Aliases: |
NEH2, NEI2 |
Location: |
Chr.8:11769663-11787346 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEIL2 | NM_001135746.1 | 5 | 1029 | NP_001129218.1 |
| NM_001135747.1 | 4 | 889 | NP_001129219.1 | |
| NM_001135748.1 | 5 | 843 | NP_001129220.1 | |
| NM_145043.2 | 5 | 1573 | NP_659480.1 | |
| XM_005272382.2 | 5 | 1005 | XP_005272439.1 | |
| XM_005272383.2 | 4 | 1301 | XP_005272440.1 | |
| XM_017013301.1 | 5 | 1446 | XP_016868790.1 | |
| AB079070.1 | 4 | 898 | BAC06478.1 | |
| AK056206.1 | 5 | 1488 | BAB71120.1 | |
| AK097389.1 | 4 | 843 | BAC05030.1 | |
| AK225837.1 | 5 | 952 | ||
| AK294224.1 | 5 | 796 | ||
| BC013952.2 | 5 | 943 | AAH13952.1 | |
| BC013964.2 | 5 | 943 | AAH13964.1 | |
| BC045822.1 | 1 | 135 | AAH45822.1 | |
| BX537529.1 | 6 | 897 | CAD97774.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616258 | Chr.8:11784657 - 11787808 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map