Assay Details
Target Gene Details
Entrez Gene ID: | 79829 |
Gene Name: | N(alpha)-acetyltransferase 40, NatD catalytic subunit |
Gene Aliases: |
NAT11, PATT1 |
Location: |
Chr.11:63938970-63957322 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA40 | NM_001300800.1 | 8 | 1740 | NP_001287729.1 |
| NM_024771.3 | 8 | 1669 | NP_079047.2 | |
| XM_006718689.3 | 8 | 1566 | XP_006718752.1 | |
| XM_011545254.2 | 9 | 1743 | XP_011543556.1 | |
| XM_011545255.2 | 9 | 1653 | XP_011543557.1 | |
| AK023910.1 | 8 | 1638 | BAB14720.1 | |
| BC041617.1 | 8 | 1598 | AAH41617.1 | |
| BC052298.1 | 8 | 1598 | AAH52298.1 | |
| BX647309.1 | 7 | 2695 | CAI46006.1 | |
| JF263566.1 | 8 | 1638 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| nsv555181 | Chr.11:63872409 - 64015880 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map