Assay Details
Target Gene Details
Entrez Gene ID: | 51026 |
Gene Name: | golgi transport 1B |
Gene Aliases: |
CGI-141, GCT2, GOT1, GOT1B, YMR292W |
Location: |
Chr.12:21501765-21518403 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLT1B | NM_016072.4 | 5 | 2802 | NP_057156.1 |
| AA910170.1 | ||||
| AB097020.1 | 5 | 2752 | BAC77373.1 | |
| AK026999.1 | 1 | 1984 | ||
| AL136571.1 | 5 | 2701 | CAB66506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv1054220 | Chr.12:21459224 - 21539169 on Build GRCh38 | Gain |
|
| nsv557743 | Chr.12:21517476 - 21535966 on Build GRCh38 | Gain |
|
| nsv528556 | Chr.12:21511945 - 21527429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map