Assay Details
Target Gene Details
Entrez Gene ID: | 28990 |
Gene Name: | asteroid homolog 1 (Drosophila) |
Gene Aliases: |
HT001 |
Location: |
Chr.3:131013873-131026854 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASTE1 | NM_001288950.1 | 3 | 698 | NP_001275879.1 |
| NM_014065.3 | 3 | 923 | NP_054784.2 | |
| NR_110229.1 | 3 | 923 | ||
| XM_017006266.1 | 3 | 839 | XP_016861755.1 | |
| XM_017006267.1 | 3 | 997 | XP_016861756.1 | |
| XM_017006268.1 | 3 | 918 | XP_016861757.1 | |
| XM_017006269.1 | 3 | 839 | XP_016861758.1 | |
| AK027258.1 | 1 | 313 | ||
| AK027806.1 | 3 | 871 | BAB55383.1 | |
| AK294156.1 | 3 | 923 | ||
| AK301388.1 | 3 | 811 | ||
| BC103709.1 | AAI03710.1 | |||
| BC110616.2 | 3 | 636 | AAI10617.1 |
More Information
Set Membership: |
|

Genomic Map