Assay Details
Target Gene Details
Entrez Gene ID: | 80018 |
Gene Name: | N(alpha)-acetyltransferase 25, NatB auxiliary subunit |
Gene Aliases: |
C12orf30, MDM20, NAP1 |
Location: |
Chr.12:112026689-112109022 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA25 | NM_024953.3 | 24 | 4063 | NP_079229.2 |
| XM_005253938.1 | 23 | 3909 | XP_005253995.1 | |
| XM_006719606.2 | 24 | 4723 | XP_006719669.1 | |
| XM_017019977.1 | 22 | 3779 | XP_016875466.1 | |
| AL832394.1 | 8 | 2131 | ||
| BX641140.1 | 23 | 5159 | CAE46062.1 | |
| BX647450.1 | 24 | 4016 |
More Information
Set Membership: |
|

Genomic Map