Assay Details
Target Gene Details
Entrez Gene ID: | 11254 |
Gene Name: | solute carrier family 6 member 14 |
Gene Aliases: |
BMIQ11 |
Location: |
Chr.X:116436579-116461458 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A14 | NM_007231.4 | NP_009162.1 | ||
| AF151978.1 | AAD49223.1 | |||
| AK304243.1 | ||||
| AK313390.1 | ||||
| BC093710.1 | AAH93710.1 | |||
| BC093712.1 | AAH93712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523582 | Chr.X:116269375 - 116491109 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map