Assay Details
Target Gene Details
Entrez Gene ID: | 9637 |
Gene Name: | fasciculation and elongation protein zeta 2 |
Gene Aliases: |
HUM3CL |
Location: |
Chr.2:36552239-36598190 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FEZ2 | NM_001042548.1 | 9 | 1238 | NP_001036013.1 |
| NM_005102.2 | 8 | 1157 | NP_005093.2 | |
| XM_006712153.2 | 9 | 1060 | XP_006712216.1 | |
| AB076184.1 | 8 | 1149 | BAD06207.1 | |
| AB209719.1 | 7 | 880 | BAD92956.1 | |
| AF113124.1 | 8 | 1056 | AAF14865.1 | |
| AK025274.1 | 7 | 837 | ||
| AK057492.1 | 1 | 1128 | ||
| AL117593.1 | 2 | 3639 | ||
| BC089390.1 | 9 | 1184 | AAH89390.1 | |
| BC110386.1 | 8 | 1112 | ||
| BX537785.1 | 9 | 1083 | CAD97842.1 | |
| U60061.1 | 5 | 510 | AAC51283.1 | |
| U69140.1 | 7 | 798 | AAB40661.1 |
More Information
Set Membership: |
|

Genomic Map