Assay Details
Target Gene Details
Entrez Gene ID: | 51126 |
Gene Name: | N(alpha)-acetyltransferase 20, NatB catalytic subunit |
Gene Aliases: |
NAT3, NAT3P, NAT5, NAT5P, dJ1002M8.1 |
Location: |
Chr.20:20017290-20033629 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA20 | NM_016100.4 | NP_057184.1 | ||
| NM_181527.3 | NP_852668.1 | |||
| NM_181528.3 | NP_852669.1 | |||
| AF085355.1 | AAD40190.1 | |||
| AK311819.1 | ||||
| AL136641.1 | CAB66576.1 | |||
| BC005181.1 | AAH05181.1 | |||
| BC008446.1 | AAH08446.1 | |||
| BG548527.1 | ||||
| BX338359.1 | ||||
| CB959658.1 |
More Information
Set Membership: |
|

Genomic Map