Assay Details
Target Gene Details
Entrez Gene ID: | 5683 |
Gene Name: | proteasome subunit alpha 2 |
Gene Aliases: |
HC3, MU, PMSA2, PSC2 |
Location: |
Chr.7:42916861-42932206 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMA2 | NM_002787.4 | 8 | 957 | NP_002778.1 |
| AK290654.1 | ||||
| BC047697.1 | 8 | 911 | AAH47697.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv606715 | Chr.7:42169735 - 42940343 on Build GRCh38 | Gain |
|
| nsv830979 | Chr.7:42806983 - 42987012 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map