Assay Details
Target Gene Details
Entrez Gene ID: | 10254 |
Gene Name: | signal transducing adaptor molecule 2 |
Gene Aliases: |
Hbp, STAM2A, STAM2B |
Location: |
Chr.2:152116801-152175992 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAM2 | NM_005843.5 | 14 | 1888 | NP_005834.4 |
| AF042273.1 | 14 | 1647 | AAC63963.1 | |
| AK023490.1 | 2 | 1110 | ||
| AK292265.1 | 14 | 1583 | ||
| AK292847.1 | 14 | 1625 | ||
| AL133600.1 | 14 | 1650 | CAB63735.1 | |
| BC028740.2 | 14 | 1888 | AAH28740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592929 | Chr.2:152076678 - 152124243 on Build GRCh38 | Loss |
|
| esv34659 | Chr.2:150858366 - 152860232 on Build GRCh38 | Loss |
|
| esv3592922 | Chr.2:151571476 - 153829714 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map