Assay Details
Target Gene Details
Entrez Gene ID: | 1585 |
Gene Name: | cytochrome P450 family 11 subfamily B member 2 |
Gene Aliases: |
ALDOS, CPN2, CYP11B, CYP11BL, CYPXIB2, P-450C18, P450C18, P450aldo |
Location: |
Chr.8:142910559-142917843 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP11B2 | NM_000498.3 | 9 | 2580 | NP_000489.3 |
| X54741.1 | 9 | 2580 | CAA38539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv991235 | Chr.8:142877756 - 142915789 on Build GRCh38 | Deletion |
|
| esv2737938 | Chr.8:142875561 - 142915781 on Build GRCh38 | Deletion |
|
| esv2661137 | Chr.8:142256294 - 142935448 on Build GRCh38 | Deletion |
|
| esv3544372 | Chr.8:142877708 - 142916066 on Build GRCh38 | Deletion |
|
| esv2673440 | Chr.8:142878652 - 142916672 on Build GRCh38 | Deletion |
|
| nsv436586 | Chr.8:142877816 - 142914663 on Build GRCh38 | Deletion |
|
| nsv470246 | Chr.8:142872807 - 142926765 on Build GRCh38 | Loss |
|
| nsv950005 | Chr.8:142910085 - 142918184 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map