Assay Details
Target Gene Details
Entrez Gene ID: | 229 |
Gene Name: | aldolase, fructose-bisphosphate B |
Gene Aliases: |
ALDB, ALDO2 |
Location: |
Chr.9:101420560-101435780 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDOB | NM_000035.3 | 3 | 324 | NP_000026.2 |
| AK026411.1 | ||||
| AK290795.1 | 3 | 318 | ||
| BC029399.1 | 3 | 318 | AAH29399.1 | |
| X01098.1 | 3 | 308 | CAA25572.1 | |
| X02747.1 | 3 | 367 | CAA26526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523287 | Chr.9:101429110 - 101440521 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map