Assay Details
Target Gene Details
Entrez Gene ID: | 27109 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B) |
Gene Aliases: |
ATPW, FB, HSU79253 |
Location: |
Chr.14:50311752-50326715 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5S | NM_001003803.2 | NP_001003803.1 | ||
| NM_001003805.2 | NP_001003805.1 | |||
| NM_015684.3 | NP_056499.2 | |||
| NR_033761.1 | ||||
| XM_005267537.3 | XP_005267594.1 | |||
| XM_011536649.2 | XP_011534951.1 | |||
| XM_011536650.2 | XP_011534952.1 | |||
| XM_011536651.2 | XP_011534953.1 | |||
| XM_011536652.1 | XP_011534954.1 | |||
| XM_011536653.2 | XP_011534955.1 | |||
| XM_011536654.2 | XP_011534956.1 | |||
| XM_011536655.2 | XP_011534957.1 | |||
| XM_011536656.2 | XP_011534958.1 | |||
| XM_011536657.2 | XP_011534959.1 | |||
| XM_017021220.1 | XP_016876709.1 | |||
| AF052186.1 | ||||
| AF390029.1 | AAL58839.1 | |||
| AK290008.1 | ||||
| AY052377.1 | AAL13058.1 | |||
| BC011549.1 | AAH11549.1 | |||
| BG699167.1 | ||||
| BI755956.1 | 1 | 541 | ||
| BU189283.1 | ||||
| U79253.1 | AAB50202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain |
|
| nsv832793 | Chr.14:50294245 - 50470857 on Build GRCh38 | Loss |
|
| nsv1052059 | Chr.14:50291535 - 50364621 on Build GRCh38 | Loss |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map