Assay Details
Target Gene Details
Entrez Gene ID: | 8930 |
Gene Name: | methyl-CpG binding domain 4, DNA glycosylase |
Gene Aliases: |
MED1 |
Location: |
Chr.3:129430944-129440179 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBD4 | NM_001276270.1 | NP_001263199.1 | ||
| NM_001276271.1 | NP_001263200.1 | |||
| NM_001276272.1 | NP_001263201.1 | |||
| NM_001276273.1 | NP_001263202.1 | |||
| NM_003925.2 | NP_003916.1 | |||
| XM_011513268.2 | XP_011511570.1 | |||
| AF072250.1 | AAC68879.1 | |||
| AF114784.1 | AAD22195.1 | |||
| AF532602.1 | AAP97338.1 | |||
| AK303013.1 | ||||
| AK312878.1 | ||||
| AM180876.1 | CAJ55826.1 | |||
| BC011752.2 | AAH11752.1 | |||
| CR450305.1 | CAG29301.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2725901 | Chr.3:129380821 - 129487939 on Build GRCh38 | Deletion |
|
| esv3597741 | Chr.3:129383049 - 129486193 on Build GRCh38 | Loss |
|
| nsv591668 | Chr.3:129403449 - 129490335 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map