Assay Details
Target Gene Details
Entrez Gene ID: | 123036 |
Gene Name: | tandem C2 domains, nuclear |
Gene Aliases: |
C14orf47, C2CD1, MTAC2D1, Tac2-N |
Location: |
Chr.14:91779751-91867536 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TC2N | NM_001128595.2 | 12 | 4414 | NP_001122067.1 |
| NM_001128596.2 | 12 | 4533 | NP_001122068.1 | |
| NM_001289134.1 | 11 | 4222 | NP_001276063.1 | |
| NM_152332.5 | 12 | 4602 | NP_689545.1 | |
| BC040503.1 | 12 | 4415 | AAH40503.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3635327 | Chr.14:91717848 - 91867474 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map